Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145139.2 | NP_660121.2 | 8 | 683 | ||
GenBank mRNA | AA038997.1 | - | 4 | 282 | ||
AK032936.1 | - | 8 | 671 | |||
AK075584.1 | - | 8 | 689 | |||
AK150895.1 | - | 8 | 678 | |||
AK151175.1 | - | 8 | 678 | |||
AK167640.1 | - | 8 | 678 | |||
AK168009.1 | - | 8 | 681 | |||
AK168295.1 | - | 8 | 680 | |||
BC021351.1 | - | 1351 | ||||
BC024463.1 | - | 8 | 683 | 4975621 |