Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sun2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205345.1 | NP_001192274.1 | 17, 18 | 2326 | ||
NM_001205346.1 | NP_001192275.1 | 17, 18 | 2320 | |||
NM_194342.3 | NP_919323.2 | 16, 17 | 2230 | |||
GenBank mRNA | AK047688.1 | - | 5, 6 | 672 | ||
AK080116.1 | - | 1, 2 | 59 | |||
AK128958.1 | - | 16, 17 | 2224 | |||
AK156246.1 | - | 17, 18 | 2327 | |||
AK171058.1 | - | 17, 18 | 2289 | |||
AK172398.1 | - | 17, 18 | 2328 | |||
AK172996.1 | - | 15, 16 | 2930 | |||
AY682987.1 | - | 16, 17 | 2086 | |||
BC023965.1 | - | 6, 7 | 878 | |||
BC027394.1 | - | 1 | 319 | |||
BC098208.1 | - | 16, 17 | 2571 | 6827666 | ||
BQ879674.1 | - | 4, 5 | 442 |