Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cerk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145475.4 | NP_663450.3 | 10, 11 | 1629 | ||
GenBank mRNA | AK042077.1 | - | 10, 11 | 1220 | ||
AK052269.1 | - | 10, 11 | 1395 | |||
AK161801.1 | - | 467 | ||||
AK169528.1 | - | 10, 11 | 1214 | |||
BC094253.1 | - | 10, 11 | 1628 | 6400872 |