Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aaas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153416.2 | NP_700465.2 | 16 | 1581 | ||
GenBank mRNA | AK034591.1 | - | 16 | 1476 | ||
AK079849.1 | - | 15 | 1589 | |||
AK083401.1 | - | 16 | 1495 | |||
AK083537.1 | - | 16 | 1502 | |||
AK087134.1 | - | 16 | 1557 | |||
AK088247.1 | - | 16 | 1498 | |||
AK134406.1 | - | 16 | 1472 | |||
AK165243.1 | - | 16 | 1496 | |||
AK166118.1 | - | 16 | 1496 | |||
AK169523.1 | - | 16 | 1533 | |||
BC018191.1 | - | 16 | 1482 | 3994248 | ||
BC025501.1 | - | 16 | 1528 | 5321013 | ||
BC026604.1 | - | 15 | 2035 |