Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146065.1 | NP_666177.1 | 4 | 302 | ||
GenBank mRNA | AK039648.1 | - | 293 | |||
AK047901.1 | - | 278 | ||||
AK154588.1 | - | 293 | ||||
AK155046.1 | - | 4 | 293 | |||
AK155444.1 | - | 4 | 320 | |||
AK157628.1 | - | 332 | ||||
BC026483.1 | - | 4 | 302 | 4503463 |