Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wiz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011717.4 | NP_035847.2 | 3 | 744 | ||
NM_212438.3 | NP_997603.3 | 2 | 397 | |||
GenBank mRNA | AB012265.1 | - | 7 | 2929 | ||
AB012266.1 | - | 3 | 438 | |||
AB255388.1 | - | 2 | 339 | |||
AK005208.1 | - | 3 | 516 | |||
BC053035.1 | - | 4 | 602 | 6400515 | ||
BU708881.1 | - | 3 | 554 | |||
CA324749.1 | - | 4 | 613 | |||
CA325151.1 | - | 4 | 603 | |||
CA327606.1 | - | 2 | 312 | |||
CB245102.1 | - | 3 | 679 | |||
CB249930.1 | - | 2 | 383 |