Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wiz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011717.4 | NP_035847.2 | 5 | 1811 | ||
NM_212438.3 | NP_997603.3 | 4 | 1464 | |||
GenBank mRNA | AB012265.1 | - | 9 | 3996 | ||
AB012266.1 | - | 5 | 1505 | |||
AB255388.1 | - | 4 | 1406 | |||
BC053035.1 | - | 7 | 1840 | 6400515 |