Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp13a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164612.1 | NP_001158084.1 | 19 | 2394 | ||
NM_001164613.1 | NP_001158085.1 | 20 | 2447 | |||
NM_172613.4 | NP_766201.3 | 19 | 2337 | |||
GenBank mRNA | AK028776.1 | - | 20 | 2449 | ||
AK029303.1 | - | 18 | 2138 | |||
AK079092.1 | - | 3 | 291 | |||
BC048410.1 | - | 19 | 2337 | 6486835 |