Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrros | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146069.4 | NP_666181.2 | 4 | 3279 | ||
GenBank mRNA | AK028367.1 | - | 2 | 2232 | ||
AK042502.1 | - | 2 | 2081 | |||
AK084858.1 | - | 4 | 3282 | |||
AK088770.1 | - | 2 | 2000 | |||
AK128897.1 | - | 3 | 2119 | |||
AK167934.1 | - | 3 | 2149 | |||
BC027411.1 | - | 3 | 2129 | 4192329 |