Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Poglut1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300827.1 | NP_001287756.1 | 4 | 624 | ||
NM_172380.4 | NP_759012.1 | 4 | 421 | |||
GenBank mRNA | AK031608.1 | - | 4 | 360 | ||
AK035948.1 | - | 4 | 369 | |||
AK036224.1 | - | 4 | 351 | |||
AK041321.1 | - | 4 | 359 | |||
AK045278.1 | - | 4 | 569 | |||
AK079886.1 | - | 4 | 390 | |||
AK167641.1 | - | 4 | 591 | |||
AK204867.1 | - | 2 | 50 | |||
BC026809.1 | - | 4 | 372 | 4205469 | ||
CB249259.1 | - | 4 | 422 |