Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C330027C09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172616.2 | NP_766204.2 | 2 | 220 | ||
GenBank mRNA | AK049356.1 | - | 2 | 222 | ||
AK160639.1 | - | 2 | 206 | |||
BC137719.1 | - | 2 | 229 | 8860738 |