Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163847.1 | NP_001157319.1 | 9 | 1825 | ||
NM_001163848.1 | NP_001157320.1 | 9 | 1911 | |||
NM_001163849.1 | NP_001157321.1 | 9 | 1825 | |||
NM_001163850.1 | NP_001157322.1 | 8 | 1807 | |||
NM_001163851.1 | NP_001157323.1 | 8 | 1893 | |||
NM_001163852.1 | NP_001157324.1 | 9 | 2043 | |||
NM_001163853.1 | NP_001157325.1 | 8 | 1871 | |||
NM_173186.4 | NP_775278.3 | 8 | 1893 | |||
GenBank mRNA | AK036088.1 | - | 9 | 1779 | ||
AK045194.1 | - | 8 | 1776 | |||
AK049306.1 | - | 8 | 1872 | |||
AK049754.1 | - | 8 | 1781 | |||
AK083833.1 | - | 9 | 1797 | |||
AK084693.1 | - | 8 | 1847 | |||
AK086293.1 | - | 8 | 1852 | |||
AK129304.1 | - | 8 | 1825 | |||
AK138431.1 | - | 9 | 1786 | |||
AK147647.1 | - | 9 | 1911 | |||
AK161054.1 | - | 9 | 2045 | |||
BC080845.1 | - | 6 | 1658 | 6850962 | ||
BC094417.1 | - | 7 | 1727 | 30943242 |