Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163847.1 | NP_001157319.1 | 8 | 1599 | ||
NM_001163848.1 | NP_001157320.1 | 8 | 1685 | |||
NM_001163849.1 | NP_001157321.1 | 8 | 1599 | |||
NM_001163850.1 | NP_001157322.1 | 7 | 1581 | |||
NM_001163851.1 | NP_001157323.1 | 7 | 1667 | |||
NM_001163852.1 | NP_001157324.1 | 8 | 1817 | |||
NM_001163853.1 | NP_001157325.1 | 7 | 1645 | |||
NM_173186.4 | NP_775278.3 | 7 | 1667 | |||
GenBank mRNA | AK036088.1 | - | 8 | 1553 | ||
AK045194.1 | - | 7 | 1549 | |||
AK049306.1 | - | 7 | 1646 | |||
AK049754.1 | - | 7 | 1555 | |||
AK083833.1 | - | 8 | 1571 | |||
AK084693.1 | - | 7 | 1621 | |||
AK086293.1 | - | 7 | 1626 | |||
AK129304.1 | - | 7 | 1599 | |||
AK138431.1 | - | 8 | 1560 | |||
AK147647.1 | - | 8 | 1685 | |||
AK161054.1 | - | 8 | 1819 | |||
BC080845.1 | - | 5 | 1432 | 6850962 | ||
BC094417.1 | - | 6 | 1501 | 30943242 |