Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207541.1 | NP_997424.1 | 4 | 1264 | ||
GenBank mRNA | AK079439.1 | - | 1 | 591 | ||
AK135853.1 | - | 4 | 1265 |