Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srsf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195485.1 | NP_001182414.1 | 2 | 199 | ||
NM_001195486.1 | NP_001182415.1 | 2 | 199 | |||
NM_001195487.1 | NP_001182416.1 | 2 | 199 | |||
NM_146083.2 | NP_666195.1 | 2 | 199 | |||
GenBank mRNA | AK034952.1 | - | 2 | 201 | ||
AK045884.1 | - | 2 | 361 | |||
AK082848.1 | - | 2 | 199 | |||
AK137795.1 | - | 2 | 200 | |||
AK161652.1 | - | 2 | 198 | |||
AK168355.1 | - | 2 | 199 | |||
BC014857.1 | - | 2 | 180 | 2646366 | ||
BC024865.1 | - | 2 | 190 | |||
BC025529.1 | - | 2 | 190 | 5342587 | ||
BC027391.1 | - | 2 | 187 | 4945988 | ||
BY740981.1 | - | 2 | 198 |