Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mib1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144860.2 | NP_659109.2 | 13 | 2565 | ||
GenBank mRNA | AK053035.1 | - | 6 | 788 | ||
AK080847.1 | - | 7 | 841 | |||
AY147848.1 | - | 13 | 2064 | |||
AY149907.1 | - | 13 | 1885 | |||
AY245539.1 | - | 13 | 2565 | |||
AY974091.1 | - | 13 | 1858 | |||
BC011287.1 | - | 2 | 162 | |||
BC083072.1 | - | 3 | 243 | |||
BC131662.1 | - | 14 | 2031 | 40110696 |