Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fhod3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289654.1 | NP_001276583.1 | 20 | 4338 | ||
NM_001289655.1 | NP_001276584.1 | 21 | 4695 | |||
NM_175276.4 | NP_780485.2 | 22 | 4791 | |||
GenBank mRNA | AB078608.1 | - | 22 | 3969 | ||
AB078609.1 | - | 20 | 3516 | |||
AB100088.1 | - | 21 | 3873 | |||
AK148245.1 | - | 20 | 3480 | |||
AK220413.1 | - | 18 | 2955 | |||
BC056371.1 | - | 12 | 2423 | |||
BC060607.1 | - | 7 | 1747 | |||
BC138046.1 | - | 20 | 3438 | 8861065 |