Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pggt1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172627.3 | NP_766215.1 | 2, 3 | 301 | ||
GenBank mRNA | AK028188.1 | - | 2, 3 | 159 | ||
AK081731.1 | - | 2, 3 | 302 | |||
AK170240.1 | - | 2, 3 | 285 | |||
BC141021.1 | - | 2, 3 | 299 | 9055633 |