Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276764.1 | NP_001263693.1 | 65 | 17231 | ||
GenBank mRNA | AK129201.1 | - | 7 | 2740 |