Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm2518 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK041085.1 | - | 1 | 230 | ||
AK077736.1 | - | 1 | 225 | |||
AK082962.1 | - | 1 | 228 | |||
AK085400.1 | - | 1 | 201 |
Gene Symbol: Taf6l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177798.1 | NP_001171269.1 | 12 | 2003 | ||
NM_146092.2 | NP_666204.2 | 12 | 1840 | |||
GenBank mRNA | AK077149.1 | - | 12 | 2004 | ||
BC020299.1 | - | 12 | 1813 | |||
BC028647.1 | - | 12 | 1787 | 1244243 |