Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ercc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011729.2 | NP_035859.2 | 9 | 2295 | ||
GenBank mRNA | AK140825.1 | - | 9 | 2124 | ||
AK160581.1 | - | 3 | 1169 | |||
AK160645.1 | - | 7 | 1667 | |||
AK164205.1 | - | 9 | 2113 | |||
BC033413.1 | - | 9 | 2015 | |||
BC053034.1 | - | 9 | 2104 | |||
BC060659.1 | - | 9 | 2094 | |||
BC092235.1 | - | 9 | 2086 | |||
BC141070.1 | - | 9 | 2125 | 9055682 | ||
D16306.1 | - | 9 | 1990 | |||
U40796.1 | - | 9 | 2191 |