Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a20b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011731.3 | NP_035861.2 | 4 | 713 | ||
GenBank mRNA | AF075261.1 | - | 4 | 537 | ||
AK076806.1 | - | 5 | 447 | |||
AK144038.1 | - | 4 | 714 | |||
AK163340.1 | - | 5 | 602 | |||
AK164706.1 | - | 4 | 558 | |||
AK164713.1 | - | 4 | 548 | |||
BC013484.1 | - | 4 | 624 | 4242168 |