Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252684.1 | NP_001239613.1 | 16 | 2246 | ||
NM_001252685.1 | NP_001239614.1 | 16 | 2075 | |||
NM_153808.2 | NP_722503.1 | 16 | 2246 | |||
GenBank mRNA | AJ534941.1 | - | 16 | 2167 | ||
AK122321.1 | - | 9 | 1192 | |||
AK143818.1 | - | 16 | 2224 | |||
AK161254.1 | - | 16 | 2076 | |||
BC007174.1 | - | 7 | 802 | 3493165 | ||
BC038345.1 | - | 16 | 2226 | 3596405 |