Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmrt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145831.3 | NP_665830.1 | 4 | 1465 | ||
GenBank mRNA | AF080623.1 | - | 436 | |||
AF539811.1 | - | 4 | 1465 | |||
AK044891.1 | - | 4 | 1466 | |||
AK140337.1 | - | 4 | 1457 | |||
AK140347.1 | - | 4 | 1460 | |||
BC027669.1 | - | 1 | 414 | 1248080 |