Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myof | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099634.1 | NP_001093104.1 | 19 | 2072 | ||
GenBank mRNA | AK086107.1 | - | 20 | 1893 | ||
AK173131.1 | - | 19 | 1853 | |||
BC044825.1 | - | 2 | 270 | |||
BC150790.1 | - | 20 | 1885 | 9087701 |