Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ablim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103177.2 | NP_001096647.1 | 7, 8 | 1061 | ||
NM_001103178.2 | NP_001096648.1 | 2, 3 | 351 | |||
NM_001290813.1 | NP_001277742.1 | 7, 8 | 1047 | |||
NM_001290815.1 | NP_001277744.1 | 7, 8 | 1061 | |||
NM_001290816.1 | NP_001277745.1 | 7, 8 | 1061 | |||
NM_178688.3 | NP_848803.3 | 7, 8 | 1047 | |||
GenBank mRNA | AF404774.1 | - | 7, 8 | 1047 | ||
AF404775.1 | - | 7, 8 | 821 | |||
AF404776.1 | - | 2, 3 | 311 | |||
AK079331.1 | - | 3, 4 | 301 | |||
AK122196.1 | - | 7, 8 | 1061 | |||
AK143907.1 | - | 7, 8 | 831 | |||
AK171860.1 | - | 958 | ||||
BC141083.1 | - | 7, 8 | 1041 | 9055695 | ||
BY268130.1 | - | 2, 3 | 336 |