Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xkr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011874.1 | NP_001011874.1 | 3 | 1970 | ||
GenBank mRNA | AK220428.1 | - | 1 | 199 | ||
AY534250.1 | - | 3 | 1970 |