Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab3gap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178690.4 | NP_848805.2 | 17 | 1917 | ||
GenBank mRNA | AK076244.1 | - | 17 | 1881 | ||
AK076399.1 | - | 17 | 1900 | |||
AK080432.1 | - | 17 | 1899 | |||
AK134604.1 | - | 17 | 1899 | |||
AK164523.1 | - | 17 | 1898 | |||
AK170768.1 | - | 17 | 1890 | |||
AK172881.1 | - | 17 | 1904 | |||
BC028996.1 | - | 1 | 325 | |||
BC038485.1 | - | 1 | 231 | |||
BC046297.1 | - | 17 | 1883 | 6486170 |