Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zranb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285945.1 | NP_001272874.1 | 19 | 2874 | ||
NM_027678.3 | NP_081954.1 | 19 | 3013 | |||
GenBank mRNA | AK050039.1 | - | 15 | 2294 | ||
AK083641.1 | - | 19 | 2992 | |||
BC066035.1 | - | 19 | 2950 | 6837116 | ||
BC117921.1 | - | 19 | 2828 | 40097463 | ||
BC117922.2 | - | 19 | 4020 | 40097464 |