Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037665.2 | NP_001032754.1 | 4 | 605 | ||
NM_011742.2 | NP_035872.2 | 4 | 563 | |||
GenBank mRNA | AK134452.1 | - | 4 | 470 | ||
AK147054.1 | - | 4 | 543 | |||
AK153830.1 | - | 4 | 547 | |||
BC025141.1 | - | 5 | 675 | |||
BC055796.1 | - | 4 | 514 | 6412519 | ||
BC062970.1 | - | 4 | 514 | 6412519 | ||
X16493.1 | - | 4 | 672 |