Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf103 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK146898.1 | - | 4 | 1698 | ||
AK168644.1 | - | 4 | 1879 | |||
BC137624.1 | - | 5 | 1236 | 8860643 | ||
D76445.1 | - | 4 | 1710 |