Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172643.5 | NP_766231.2 | 2 | 1062 | ||
GenBank mRNA | AK034756.1 | - | 1 | 161 | ||
AK036544.1 | - | 2 | 1061 | |||
AK163302.1 | - | 1 | 335 | |||
BC046465.1 | - | 1 | 231 | |||
BC150859.1 | - | 1 | 401 | 9087770 | ||
BC172012.1 | - | 1 | 401 |