Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172462.4 | NP_766050.1 | 2, 3 | 131 | ||
GenBank mRNA | AK032872.1 | - | 2, 3 | 145 | ||
AK085077.1 | - | 1, 2 | 117 | |||
BB623389.1 | - | 2, 3 | 145 | |||
BC058553.1 | - | 2, 3 | 129 | |||
BC094590.1 | - | 1, 2 | 116 | 30616053 | ||
BC139312.1 | - | 1, 2 | 111 | 8862334 | ||
BC139315.1 | - | 1, 2 | 111 | 8862337 |