Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172644.3 | NP_766232.1 | 10 | 1426 | ||
GenBank mRNA | AK030952.1 | - | 10 | 1415 | ||
AK033634.1 | - | 9 | 2249 | |||
AK037337.1 | - | 10 | 1445 | |||
AK048454.1 | - | 10 | 1403 | |||
AK138809.1 | - | 10 | 1427 | |||
AK154348.1 | - | 10 | 1454 | |||
AK164885.1 | - | 10 | 1396 | |||
BC046429.1 | - | 3 | 191 | |||
BC057943.1 | - | 10 | 1450 | |||
BC076606.1 | - | 10 | 1418 | 30625515 | ||
BC089191.1 | - | 10 | 1400 | 30473852 |