Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039482.1 | NP_001034571.1 | 5, 6 | 1016 | ||
GenBank mRNA | AK032131.1 | - | 3, 4 | 503 | ||
AK053102.1 | - | 5, 6 | 966 | |||
AK054169.1 | - | 5, 6 | 984 | |||
AK220547.1 | - | 5, 6 | 995 | |||
BC019571.1 | - | 5, 6 | 988 | 4235202 | ||
BC060183.1 | - | 5, 6 | 945 |