Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmo4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144878.1 | NP_659127.1 | 7 | 991 | ||
GenBank mRNA | AF461145.1 | - | 7 | 991 | ||
BC106858.1 | - | 6 | 982 | 40047236 | ||
BC106859.1 | - | 6 | 982 | 40047237 |