Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tiprl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145513.4 | NP_663488.1 | 7 | 924 | ||
GenBank mRNA | AK049119.1 | - | 7 | 872 | ||
AK049784.1 | - | 7 | 910 | |||
AK075774.1 | - | 7 | 904 | |||
AK208286.1 | - | 2 | 142 | |||
BC058250.1 | - | 7 | 863 | 6315314 |