Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp185 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109043.1 | NP_001102513.1 | 16 | 1438 | ||
NM_009549.3 | NP_033575.3 | 16 | 1441 | |||
GenBank mRNA | AK014497.1 | - | 5 | 489 | ||
AK048188.1 | - | 16 | 1419 | |||
BC039772.1 | - | 13 | 1026 | 4191053 | ||
EF494744.1 | - | 16 | 1421 | |||
U46687.1 | - | 15 | 1267 |