Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfap2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153154.2 | NP_694794.1 | 7 | 1332 | ||
GenBank mRNA | AF421891.1 | - | 7 | 1359 | ||
AK132112.1 | - | 7 | 1334 | |||
BC152324.1 | - | 7 | 1281 | 40141049 | ||
BC152325.1 | - | 7 | 1281 | 40141051 |