Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnq5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160139.1 | NP_001153611.1 | 15 | 2699 | ||
NM_023872.3 | NP_076361.1 | 14 | 2642 | |||
GenBank mRNA | AF263836.1 | - | 14 | 2117 | ||
AK033079.1 | - | 8 | 1200 | |||
AK147264.1 | - | 15 | 2702 | |||
AY679158.1 | - | 14 | 2281 |