Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290350.1 | NP_001277279.1 | 4 | 557 | ||
NM_001290351.1 | NP_001277280.1 | 4 | 548 | |||
NM_001290353.1 | NP_001277282.1 | 4 | 455 | |||
NM_009554.4 | NP_033580.3 | 4 | 545 | |||
GenBank mRNA | AK032469.1 | - | 4 | 427 | ||
BC024863.1 | - | 2 | 168 | |||
BC063757.1 | - | 4 | 429 | 6331110 | ||
CF915126.1 | - | 4 | 455 | |||
CO430987.1 | - | 4 | 427 | |||
CO434318.1 | - | 4 | 382 | |||
X64413.1 | - | 4 | 351 | |||
X89264.1 | - | 5 | 457 |