Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290350.1 | NP_001277279.1 | 4 | 688 | ||
NM_001290351.1 | NP_001277280.1 | 4 | 679 | |||
NM_001290353.1 | NP_001277282.1 | 4 | 586 | |||
NM_009554.4 | NP_033580.3 | 4 | 676 | |||
GenBank mRNA | AK032469.1 | - | 4 | 558 | ||
BC024863.1 | - | 2 | 299 | |||
BC063757.1 | - | 4 | 560 | 6331110 | ||
CF915126.1 | - | 4 | 586 | |||
CO430987.1 | - | 4 | 558 | |||
CO434318.1 | - | 4 | 513 |