Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183019.2 | NP_898840.2 | 10 | 1669 | ||
GenBank mRNA | AK034013.1 | - | 10 | 1633 | ||
AK122448.2 | - | 6 | 724 | |||
AK134629.1 | - | 1632 | ||||
AK138598.1 | - | 10 | 1629 | |||
AK163424.1 | - | 10 | 1651 | |||
AK169957.1 | - | 10 | 1689 | |||
BC055804.1 | - | 10 | 1630 | |||
BC094608.1 | - | 5189 | ||||
BC139371.1 | - | 10 | 1652 | 8862393 | ||
BC139397.1 | - | 10 | 1652 | 8862419 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK050761.1 | - | 197 | Chr. - - [-] on Build |