Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf3c3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033194.2 | NP_001028366.1 | 11 | 1514 | ||
GenBank mRNA | AK145040.1 | - | 1516 | |||
AK160997.1 | - | 11 | 1502 | |||
AK165830.1 | - | 11 | 1515 | |||
BC141157.1 | - | 11 | 1504 | 9055769 |