Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abca12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175210.3 | NP_780419.2 | 44 | 6794 | ||
GenBank mRNA | BC158053.1 | - | 44 | 6607 | 9088162 | |
BC158063.1 | - | 44 | 6607 | 9007362 |