Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dner | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152915.1 | NP_690879.1 | 11 | 2232 | ||
GenBank mRNA | AB067650.1 | - | 11 | 1881 | ||
AF370126.1 | - | 11 | 1888 | |||
AK044597.1 | - | 11 | 1995 | |||
AK152755.1 | - | 6 | 635 | |||
AK153235.1 | - | 6 | 635 | |||
AY032924.1 | - | 11 | 1888 | |||
BC022636.1 | - | 6 | 695 | |||
BC034634.1 | - | 11 | 2232 | 5365190 |