Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco4c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172658.3 | NP_766246.1 | 12 | 2000 | ||
GenBank mRNA | AK049253.1 | - | 12 | 2000 | ||
AK049944.1 | - | 1965 | ||||
AY654588.1 | - | 12 | 2000 | |||
BC119022.1 | - | 12 | 1959 | 8733771 |