Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppip5k2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173760.5 | NP_776121.4 | 18, 19 | 2574 | ||
GenBank mRNA | AK087712.1 | - | 18, 19 | 2504 | ||
AK129140.1 | - | 18, 19 | 2510 | |||
AK138622.1 | - | 18, 19 | 2891 | |||
AK149804.1 | - | 18, 19 | 2506 | |||
AK162179.1 | - | 11, 12 | 1293 | |||
BC046411.1 | - | 8, 9 | 887 | |||
BC053396.1 | - | 17, 18 | 2076 | 30044788 |