Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camk1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290374.1 | NP_001277303.1 | 3 | 838 | ||
NM_001290375.1 | NP_001277304.1 | 4 | 913 | |||
NM_001290376.1 | NP_001277305.1 | 4 | 429 | |||
NM_177343.4 | NP_796317.2 | 4 | 913 | |||
GenBank mRNA | AK052401.1 | - | 4 | 334 | ||
AK053173.1 | - | 4 | 667 | |||
AK147616.1 | - | 4 | 914 | |||
AK147657.1 | - | 4 | 624 | |||
AK154434.1 | - | 4 | 502 | |||
AK162754.1 | - | 4 | 628 | |||
AK170123.1 | - | 3 | 597 | |||
AK170777.1 | - | 4 | 678 | |||
AY273822.1 | - | 4 | 429 | |||
BC141413.1 | - | 4 | 413 | 9056026 | ||
BC141414.1 | - | 4 | 413 | 9056027 |