Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zkscan5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167944.1 | NP_001161416.1 | 1 | 1040 | ||
NM_016683.2 | NP_057892.2 | 1 | 1040 | |||
GenBank mRNA | AB221634.1 | - | 1 | 304 | ||
AK019484.1 | - | 1 | 1031 | |||
AK173086.1 | - | 1 | 1035 | |||
BC060248.1 | - | 1 | 935 | 5715760 | ||
U62907.1 | - | 1 | 364 |